Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab9b
Entrez Gene ID
367915
Gene Name
RAB9B, member RAS oncogene family
Gene Aliases
RGD1562951
Chromosome Location
Chr.X: 107531404 - 107542510 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109018.1 NP_001102488.1 1 - 2 147 141

View Full Product Details