Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Colec10
Entrez Gene ID
299928
Gene Name
collectin sub-family member 10 (C-type lectin)
Gene Aliases
-
Chromosome Location
Chr.7: 93975451 - 94035999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130541.1 NP_001124013.1 4 - 5 355 89

View Full Product Details