The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108529.1 | NP_001101999.1 | 4 - 5 | 516 | 77 |
XM_006256301.2 | XP_006256363.1 | 4 - 5 | 527 | 77 |