The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017469.1 | NP_001017469.1 | 5 - 6 | 1069 | 135 |
GenBank mRNA | BC078980.1 | - | 5 - 6 | 1069 | 135 |