Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcap29
Entrez Gene ID
298943
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
-
Chromosome Location
Chr.6: 50883758 - 50923743 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006980.1 NP_001006981.1 3 - 4 378 107
XM_006239982.2 XP_006240044.1 4 - 5 499 107
GenBank mRNA BC083576.1 - 3 - 4 378 107

View Full Product Details