Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcd10
Entrez Gene ID
494345
Gene Name
programmed cell death 10
Gene Aliases
-
Chromosome Location
Chr.2: 173966701 - 174012729 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009542.2 NP_001009542.1 6 - 7 674 159
XM_006232501.2 XP_006232563.1 6 - 7 635 159
GenBank mRNA BC067245.1 - 6 - 7 678 159
FQ213019.1 - 6 - 7 675 159

View Full Product Details