Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mab21l1
Entrez Gene ID
688394
Gene Name
mab-21-like 1 (C. elegans)
Gene Aliases
-
Chromosome Location
Chr.2: 145174876 - 145177265 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109497.1 NP_001102967.1 1 - 1 2063 117

View Full Product Details