Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtp1
Entrez Gene ID
288006
Gene Name
receptor (chemosensory) transporter protein 1
Gene Aliases
RGD1560980
Chromosome Location
Chr.11: 80823989 - 80826505 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105870.1 NP_001099340.1 1 - 2 290 133

View Full Product Details