The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198735.2 | NP_942030.1 | 2 - 3 | 776 | 81 |
XM_008759695.1 | XP_008757917.1 | 2 - 3 | 1004 | 81 | |
XM_008759698.1 | XP_008757920.1 | 5 - 6 | 1365 | 81 | |
GenBank mRNA | BC099070.1 | - | 2 - 3 | 776 | 81 |
FQ227811.1 | - | 1 - 2 | 550 | 81 | |
FQ230306.1 | - | 4 - 5 | 1112 | 81 | |
M85193.1 | - | 2 - 3 | 81 |