Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab15
Entrez Gene ID
299156
Gene Name
RAB15, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.6: 99845952 - 99870320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198749.2 NP_942044.1 5 - 6 786 96
GenBank mRNA BC094954.1 - 5 - 6 786 96
M83679.1 - 5 - 6 630 96

View Full Product Details