The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017297.2 | NP_058993.1 | 1 - 2 | 180 | 87 |
GenBank mRNA | BC087022.1 | - | 1 - 2 | 180 | 87 |
D50135.1 | - | 1 - 2 | 133 | 87 | |
L35771.1 | - | 1 - 2 | 185 | 87 |