Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slamf9
Entrez Gene ID
289235
Gene Name
SLAM family member 9
Gene Aliases
-
Chromosome Location
Chr.13: 90836420 - 90839200 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105971.1 NP_001099441.1 4 - 4 1079 131
GenBank mRNA FQ225404.1 - 4 - 4 1040 131
FQ232537.1 - 4 - 4 1040 131
FQ233573.1 - 4 - 4 1040 131
FQ235209.1 - 4 - 4 1065 131

View Full Product Details