The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013231.1 | NP_001013249.1 | 4 - 4 | 2261 | 135 |
GenBank mRNA | AJ243949.1 | - | 4 - 4 | 2303 | 135 |
BC085766.1 | - | 4 - 4 | 2261 | 135 | |
FQ235065.1 | - | 1 - 1 | 1376 | 135 |