Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srsf9
Entrez Gene ID
288701
Gene Name
serine/arginine-rich splicing factor 9
Gene Aliases
Sfrs9
Chromosome Location
Chr.12: 47039714 - 47046022 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009255.1 NP_001009255.1 4 - 4 785 128
GenBank mRNA BC087684.1 - 4 - 4 785 128

View Full Product Details