Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps26b
Entrez Gene ID
300472
Gene Name
vacuolar protein sorting 26 homolog B (S. pombe)
Gene Aliases
-
Chromosome Location
Chr.8: 28054407 - 28075514 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106809.1 NP_001100279.1 6 - 6 3449 72
GenBank mRNA CB578687.1 - 1 - 1 494 72

View Full Product Details