The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022686.2 | NP_073177.1 | 2 - 2 | 491 | 154 |
GenBank mRNA | AY936209.1 | - | 2 - 2 | 491 | 154 |
BC100619.1 | - | 2 - 2 | 495 | 154 |