Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lhfp
Entrez Gene ID
499615
Gene Name
lipoma HMGIC fusion partner
Gene Aliases
RGD1560177
Chromosome Location
Chr.2: 142262195 - 142461980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109183.1 NP_001102653.1 4 - 4 1909 142
GenBank mRNA BC091355.1 - 3 - 3 1644 142

View Full Product Details