The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053321.2 | NP_445773.1 | 2 - 2 | 3256 | 114 |
XM_006239063.2 | XP_006239125.1 | 2 - 2 | 3581 | 114 | |
XM_006239064.2 | XP_006239126.1 | 2 - 2 | 3867 | 114 | |
GenBank mRNA | BC072491.1 | - | 2 - 2 | 3256 | 114 |