Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt34
Entrez Gene ID
303528
Gene Name
keratin 34
Gene Aliases
Ka29, Krt1-4
Chromosome Location
Chr.10: 87950364 - 87954055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008758.1 NP_001008758.1 1 - 2 355 141

View Full Product Details