Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt84
Entrez Gene ID
315320
Gene Name
keratin 84, type II
Gene Aliases
Kb24
Chromosome Location
Chr.7: 143180229 - 143187663 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008812.1 NP_001008812.1 7 - 8 1461 148

View Full Product Details