Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ugt2b
- Entrez Gene ID
-
24862
- Gene Name
- UDP glycosyltransferase 2 family, polypeptide B
- Gene Aliases
-
Ugt2b2
- Chromosome Location
-
-
- Gene Symbol
- LOC100364155
- Entrez Gene ID
-
100364155
- Gene Name
- hypothetical LOC100364155
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 22326004 - 22346288 on Build Rnor_6.0
- Gene Symbol
- Ugt2b17
- Entrez Gene ID
-
266685
- Gene Name
- UDP glucuronosyltransferase 2 family, polypeptide B15
- Gene Aliases
-
Rlug38, UDPGT 2B5, Udpgt, Udpgtr-3, Ugt2b3, Ugt2b5
- Chromosome Location
-
Chr.14: 22374877 - 22486291 on Build Rnor_6.0
View Full Product Details