Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ervfrd-1
Entrez Gene ID
290348
Gene Name
endogenous retrovirus group FRD, member 1
Gene Aliases
Herv-frd, RGD1566115
Chromosome Location
Chr.15: 51174989 - 51189948 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024239.1 NP_001019410.1 1 - 1 1795 101

View Full Product Details