Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Serpinb3
Entrez Gene ID
304688
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 3
Gene Aliases
-
Chromosome Location
Chr.13: 27186701 - 27193210 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008887.1 NP_001008887.1 1 - 1 129 155

View Full Product Details