The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037850.1 | NP_001032939.1 | 2 - 3 | 210 | 146 |
NM_001037852.1 | NP_001032941.1 | 2 - 3 | 270 | 146 | |
GenBank mRNA | DQ012092.1 | - | 2 - 3 | 210 | 146 |
DQ012093.1 | - | 2 - 3 | 270 | 146 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_002728419.4 | XP_002728465.2 | 2 - 3 | 397 | 146 |
XM_006253377.2 | XP_006253439.1 | 2 - 3 | 400 | 146 | |
GenBank mRNA | AY600145.1 | - | 2 - 3 | 230 | 146 |