The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022617.1 | NP_072139.1 | 1 - 1 | 1467 | 113 |
XM_006231115.2 | XP_006231177.1 | 2 - 2 | 1544 | 113 | |
XM_008760261.1 | XP_008758483.1 | 2 - 2 | 1636 | 113 | |
GenBank mRNA | AF156540.1 | - | 1 - 1 | 1467 | 113 |