Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r12
Entrez Gene ID
690566
Gene Name
vomeronasal 2 receptor, 12
Gene Aliases
-
Chromosome Location
Chr.1: 64849657 - 64950897 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099488.1 NP_001092958.1 2 - 3 488 132

View Full Product Details