The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079712.1 | NP_001073180.1 | - | 39 | 79 |
GenBank mRNA | BC128760.1 | - | 1 - 2 | 39 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003750432.1 | XP_003750480.1 | 1 - 2 | 438 | 79 |