Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc35g3
Entrez Gene ID
691976
Gene Name
solute carrier family 35, member G3
Gene Aliases
Amac1
Chromosome Location
Chr.10: 56366578 - 56368360 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127658.1 NP_001121130.1 1 - 2 41 75
GenBank mRNA BC159402.1 - 1 - 2 41 75

View Full Product Details