Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sry
- Entrez Gene ID
-
25221
- Gene Name
- sex determining region Y
- Gene Aliases
-
SRYGENE, Sry1, Sry3BI, Tdf, Tdy
- Chromosome Location
-
Chr.Y: 327176 - 327685 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012772.1 |
NP_036904.1 |
1 - 1 |
352 |
102 |
- Gene Symbol
- LOC103694545
- Entrez Gene ID
-
103694545
- Gene Name
- sex-determining region Y protein-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 406648 - 408636 on Build Rnor_6.0
- Gene Symbol
- LOC103694550
- Entrez Gene ID
-
103694550
- Gene Name
- sex-determining region Y protein-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 472964 - 474988 on Build Rnor_6.0
- Gene Symbol
- LOC103694554
- Entrez Gene ID
-
103694554
- Gene Name
- sex-determining region Y protein-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 644648 - 646381 on Build Rnor_6.0
- Gene Symbol
- LOC103694555
- Entrez Gene ID
-
103694555
- Gene Name
- sex-determining region Y protein-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 704356 - 706089 on Build Rnor_6.0
View Full Product Details