Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scube3
Entrez Gene ID
294297
Gene Name
signal peptide, CUB domain, EGF-like 3
Gene Aliases
-
Chromosome Location
Chr.20: 7718260 - 7749841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271362.1 NP_001258291.1 19 - 20 2607 101
XM_008772802.1 XP_008771024.1 19 - 20 4620 101
XM_008772803.1 XP_008771025.1 20 - 21 2677 101
XM_008772804.1 XP_008771026.1 18 - 19 4572 101
XM_008772805.1 XP_008771027.1 19 - 20 4620 101
XM_008772806.1 XP_008771028.1 18 - 19 4572 101

View Full Product Details