Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sult1b1
Entrez Gene ID
64305
Gene Name
sulfotransferase family, cytosolic, 1B, member 1
Gene Aliases
ST1B1
Chromosome Location
Chr.14: 22142412 - 22155246 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022513.2 NP_071958.1 2 - 3 362 99
XM_008770056.1 XP_008768278.1 2 - 3 236 99
GenBank mRNA D89375.1 - 2 - 3 313 99
FQ209476.1 - 2 - 3 331 99
FQ209486.1 - 2 - 3 396 99
FQ209579.1 - 2 - 3 417 99
FQ210801.1 - 2 - 3 404 99
FQ212608.1 - 2 - 3 373 99
FQ218295.1 - 2 - 3 375 99
U38419.1 - 2 - 3 365 99

View Full Product Details