Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAW1
Entrez Gene ID
851206
Gene Name
Gene Aliases
YAL027W
Chromosome Location
Chr.I: 94687 - 95472 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178172.1 NP_009375.1 1 - 1 546 106

View Full Product Details