Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACH1
Entrez Gene ID
852266
Gene Name
Gene Aliases
YBL015W
Chromosome Location
Chr.II: 194122 - 195702 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178255.1 NP_009538.1 1 - 1 1055 94

View Full Product Details