Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC17
Entrez Gene ID
852230
Gene Name
Gene Aliases
RNS3, YBL050W
Chromosome Location
Chr.II: 125125 - 126119 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178290.1 NP_009503.1 1 - 2 32 97

View Full Product Details