Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGA2
Entrez Gene ID
852291
Gene Name
Gene Aliases
UGA5, YBR006W
Chromosome Location
Chr.II: 247010 - 248503 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178354.1 NP_009560.1 1 - 1 1345 90

View Full Product Details