Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKL1
Entrez Gene ID
852351
Gene Name
Gene Aliases
YBR059C
Chromosome Location
Chr.II: 356861 - 360187 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178407.1 NP_009615.1 1 - 1 3245 78

View Full Product Details