Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFA38
Entrez Gene ID
852456
Gene Name
Gene Aliases
YBR159W
Chromosome Location
Chr.II: 558685 - 559728 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178507.1 NP_009717.1 1 - 1 852 90

View Full Product Details