Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSM1
Entrez Gene ID
850447
Gene Name
Gene Aliases
YCR086W
Chromosome Location
Chr.III: 263392 - 263964 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178792.1 NP_010009.1 1 - 1 452 95

View Full Product Details