Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTH1
Entrez Gene ID
851564
Gene Name
Gene Aliases
YDR001C
Chromosome Location
Chr.IV: 450220 - 452475 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180309.1 NP_010284.1 1 - 1 2049 98

View Full Product Details