Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NBP2
Entrez Gene ID
851740
Gene Name
Gene Aliases
YDR162C
Chromosome Location
Chr.IV: 780390 - 781100 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180469.3 NP_010446.3 1 - 1 545 73

View Full Product Details