Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPT1
Entrez Gene ID
852008
Gene Name
Gene Aliases
BRA6, YDR399W
Chromosome Location
Chr.IV: 1270068 - 1270733 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180707.3 NP_010687.3 1 - 1 552 72

View Full Product Details