Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGO1
Entrez Gene ID
852081
Gene Name
Gene Aliases
YDR470C
Chromosome Location
Chr.IV: 1399706 - 1401214 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180778.2 NP_010758.3 1 - 1 1447 92

View Full Product Details