Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGD2
Entrez Gene ID
850497
Gene Name
Gene Aliases
YFL047W
Chromosome Location
Chr.VI: 40421 - 42565 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179920.1 NP_428268.1 1 - 1 1952 95

View Full Product Details