Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCW11
Entrez Gene ID
852856
Gene Name
Gene Aliases
YGL028C
Chromosome Location
Chr.VII: 441281 - 442909 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180893.3 NP_011487.3 1 - 1 1550 99

View Full Product Details