Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKI8
Entrez Gene ID
852659
Gene Name
Gene Aliases
REC103, YGL213C
Chromosome Location
Chr.VII: 90054 - 91247 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181078.1 NP_011302.1 1 - 1 847 66

View Full Product Details