Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS29
Entrez Gene ID
856403
Gene Name
Gene Aliases
PEP11, VPT6, YHR012W
Chromosome Location
Chr.VIII: 129481 - 130448 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179142.1 NP_011876.1 1 - 2 41 103

View Full Product Details