Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARP1
Entrez Gene ID
856530
Gene Name
Gene Aliases
ACT5, YHR129C
Chromosome Location
Chr.VIII: 362999 - 364153 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179259.1 NP_011997.1 1 - 1 951 72

View Full Product Details