Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TES1
Entrez Gene ID
853477
Gene Name
Gene Aliases
PTE1, YJR019C
Chromosome Location
Chr.X: 467232 - 468281 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181677.3 NP_012553.3 1 - 1 953 52

View Full Product Details