Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAM1
Entrez Gene ID
853532
Gene Name
Gene Aliases
YJR069C
Chromosome Location
Chr.X: 568806 - 569399 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181727.1 NP_012603.1 1 - 1 452 102

View Full Product Details