Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUD2
Entrez Gene ID
853788
Gene Name
Gene Aliases
YKL074C
Chromosome Location
Chr.XI: 294610 - 296193 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179640.1 NP_012849.1 1 - 1 1351 101

View Full Product Details